2-Hydroxy-6-methylpyridine, 98%
2-Hydroxy-6-methylpyridine, 98%
2-Hydroxy-6-methylpyridine, 98%
2-Hydroxy-6-methylpyridine, 98%
Thermo Scientific Chemicals

2-Hydroxy-6-methylpyridine, 98%

CAS: 3279-76-3 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14764.14
also known as A14764-14
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS3279-76-3
IUPAC Name6-methyl-1,2-dihydropyridin-2-one
Molecular FormulaC6H7NO
InChI KeyJEAVIRYCMBDJIU-UHFFFAOYSA-N
SMILESCC1=CC=CC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
Assay (HPLC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only