Methyl 3-aminobenzoate, 98%
Methyl 3-aminobenzoate, 98%
Methyl 3-aminobenzoate, 98%
Methyl 3-aminobenzoate, 98%
Thermo Scientific Chemicals

Methyl 3-aminobenzoate, 98%

CAS: 4518-10-9 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14766.14
also known as A14766-14
Price (USD)
178.65
Special offer
Online exclusive
Ends: 30-Jun-2026
210.00
Save 31.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
178.65
Special offer
Online exclusive
Ends: 30-Jun-2026
210.00
Save 31.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS4518-10-9
IUPAC Namemethyl 3-aminobenzoate
Molecular FormulaC8H9NO2
InChI KeyVZDNXXPBYLGWOS-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to pale brown to brown
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Methyl 3-aminobenzoate, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-aminobenzoate, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water (0.1-1%) .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. PA Hepworth.; J McCombie.; JP Simons. High resolution electronic spectroscopy of molecular conformers. Methyl-and ethyl-3-aminobenzoic acid esters. Chemical physics.1996, 249 (5-6), 341-350.
  2. J Li, Y Sha. A convenient synthesis of amino acid methyl esters. Molecules.2008, 13 (5), 1111-1119.