HEPES, 99%
HEPES, 99%
HEPES, 99%
Thermo Scientific Chemicals

HEPES, 99%

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14777.18
also known as A14777-18
Price (USD)
57.65
Online Exclusive
63.90
Save 6.25 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
63.90
Save 6.25 (10%)
Each
Chemical Identifiers
CAS7365-45-9
IUPAC Name2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular FormulaC8H18N2O4S
InChI KeyJKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILESOCCN1CCN(CCS(O)(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
View more
HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.  
RUO – Research Use Only

General References:

  1. Maus, S.; Breeman, W.; Buchholz, H. G.; de Blois, E.; Schreckenberger, M. Simple and reliable chromatographic technique for the determination of HEPES in Ga-68-labeled pharmaceuticals. J. Nucl. Med. 2015, 56, 1168-1168.
  2. Taha, M.; Khoiroh, I.; Lee, M. J. Phase behavior and molecular dynamics simulation studies of new aqueous two-phase separation systems induced by HEPES buffer. J. Phys. Chem. B 2013, 117, (2), 563-582.