2-Methylbenzophenone, 98+%
2-Methylbenzophenone, 98+%
2-Methylbenzophenone, 98+%
Thermo Scientific Chemicals

2-Methylbenzophenone, 98+%

CAS: 131-58-8 | C14H12O | 196.249 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14779.09
also known as A14779-09
Price (USD)
44.65
Online Exclusive
49.00
Save 4.35 (9%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
44.65
Online Exclusive
49.00
Save 4.35 (9%)
Each
Chemical Identifiers
CAS131-58-8
IUPAC Name(2-methylphenyl)(phenyl)methanone
Molecular FormulaC14H12O
InChI KeyCKGKXGQVRVAKEA-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5930-1.5970 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.0%
2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperature and pressure.
RUO – Research Use Only

General References:

  1. PK Das.; JC Scaiano. The formation of two photoenols via a common pathway in the photochemistry of 2-methylbenzophenone. Journal of Photochemistry.1980, 12 (1), 85-90.
  2. T Nakayama.; K Hamanoue.; T Hidaka.; M Okamoto. Photoenolization of 2-methylbenzophenone studied by picosecond and nanosecond laser spectroscopy. Journal of Photochemistry.1980, 24 (1), 71-78.
  3. For a review of the Elbs reaction, in which 2-alkylbenzophenones are cyclized at high temperatures to anthracenes, see: Org. React., 1, 129 (1942).