2-Chlorobenzoyl chloride, 97%
2-Chlorobenzoyl chloride, 97%
2-Chlorobenzoyl chloride, 97%
Thermo Scientific Chemicals

2-Chlorobenzoyl chloride, 97%

CAS: 609-65-4 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1000 g
Catalog number A14785.0B
also known as A14785-0B
Price (USD)
340.65
Online Exclusive
378.00
Save 37.35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
340.65
Online Exclusive
378.00
Save 37.35 (10%)
Each
Chemical Identifiers
CAS609-65-4
IUPAC Name2-chlorobenzoyl chloride
Molecular FormulaC7H4Cl2O
InChI KeyONIKNECPXCLUHT-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5705-1.5755 @ 20?C
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to pale yellow
2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.

Solubility
Soluble in acetone, ether, and alcohol. Solubility in water it decomposes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong bases, alcohols, oxidizing agents.
RUO – Research Use Only

General References:

  1. Y Zhang.; T Wei.; L Wang. Phase-transfer-catalyzed synthesis of N-aryl-N'-(2-chlorobenzoyl)-thiourea derivatives. Synthetic communications. 1997, 27 (5), 751-756.
  2. PC Reeves. Carboxylation of aromatic compounds: ferrocenecarboxylic acid. Organic Syntheses. 1977, 1 (5), 51-56.
  3. Can be used to introduce a carboxyl group into an aromatic ring by Friedel-Crafts acylation, followed by cleavage of the resulting ketone with KO-t-Bu. With this aryl group, cleavage occurs in only one of the two possible senses. For an example, (ferrocenecarboxylic acid), see: Org. Synth. Coll., 6, 625 (1988).