Xanthone, 99%
Xanthone, 99%
Xanthone, 99%
Thermo Scientific Chemicals

Xanthone, 99%

CAS: 90-47-1 | C13H8O2 | 196.205 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A14812.22
also known as A14812-22
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS90-47-1
IUPAC Name9H-xanthen-9-one
Molecular FormulaC13H8O2
InChI KeyJNELGWHKGNBSMD-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2OC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or pale grey or pale pink
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)173-179°C
Xanthone is used predominantly as oxygenated polycyclic aromatic compounds, and as a vasorelaxant. It can be used as a fluorescent agent, as well as an intermediate in organic synthesis and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xanthone is used predominantly as oxygenated polycyclic aromatic compounds, and as a vasorelaxant. It can be used as a fluorescent agent, as well as an intermediate in organic synthesis and chemical research.

Solubility
It is Soluble in water (partly), chloroform, hot Toluene, ether (slightly), and hot alcohol.

Notes
Stable under normal conditions. Incompatible to strong oxidizing agents.
RUO – Research Use Only

General References:

  1. J. C. Scaiano. Solvent effects in the photochemistry of xanthone. J. Org. Chem. 1980, 102, (26), 7747-7753.
  2. CC Chen.; et.al. Garcinone E, a xanthone derivative, has potent cytotoxic effect against hepatocellular carcinoma cell lines. Biochimica et Biophysica Acta (BBA) - General Subjects. 2002, 68, (3), 975-979.