2-Chloro-5-nitrobenzaldehyde, 97%
2-Chloro-5-nitrobenzaldehyde, 97%
2-Chloro-5-nitrobenzaldehyde, 97%
2-Chloro-5-nitrobenzaldehyde, 97%
Thermo Scientific Chemicals

2-Chloro-5-nitrobenzaldehyde, 97%

CAS: 6361-21-3 | C7H4ClNO3 | 185.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A14824.09
also known as A14824-09
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.70
Save 7.05 (16%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.70
Save 7.05 (16%)
Each
Add to cart
Chemical Identifiers
CAS6361-21-3
IUPAC Name2-chloro-5-nitrobenzaldehyde
Molecular FormulaC7H4ClNO3
InChI KeyVFVHWCKUHAEDMY-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Cl)C(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1.5%
Melting Point (clear melt)74.0-84.0°C
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White to cream to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only