p-Terphenyl, 99+%
p-Terphenyl, 99+%
p-Terphenyl, 99+%
Thermo Scientific Chemicals

p-Terphenyl, 99+%

CAS: 92-94-4 | C18H14 | 230.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
Catalog number A14833.22
also known as A14833-22
Price (USD)
106.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
106.00
Each
Chemical Identifiers
CAS92-94-4
IUPAC Name4-phenyl-1,1'-biphenyl
Molecular FormulaC18H14
InChI KeyXJKSTNDFUHDPQJ-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (HPLC)≥99.0%
Identification (FTIR)Conforms
Melting Point211.0-217.0°C
p-Terphenyl is a specialty material, and may be used in ionized radiation detectors, nonpolar laser dye, and single molecule optical probe of scanning near-field microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Terphenyl is a specialty material, and may be used in ionized radiation detectors, nonpolar laser dye, and single molecule optical probe of scanning near-field microscopy.

Solubility
Insoluble in water. Soluble in hot benzene. Very soluble in hot ethyl alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Boens N.; Qin W, Basarić N.; Hofkens J.; Ameloot M.; Pouget J.; Lefèvre JP.etal. Fluorescence lifetime standards for time and frequency domain fluorescence spectroscopy. Analytical chemistry . 2007, 79, (3), 2137-2149 .
  2. JK Liu .; L Hu .; ZJ Dong .; Q Hu. DPPH Radical Scavenging Activity of Ten Natural p-Terphenyl Derivatives Obtained from Three Edible Mushrooms Indigenous to China. Chemistry & biodiversity. 2004, 1, (4), 601-605.