2-Chloroacetamidine hydrochloride, 96%
2-Chloroacetamidine hydrochloride, 96%
2-Chloroacetamidine hydrochloride, 96%
Thermo Scientific Chemicals

2-Chloroacetamidine hydrochloride, 96%

CAS: 10300-69-3 | C2H6Cl2N2 | 128.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14835.06
also known as A14835-06
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS10300-69-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown to brown to black
FormPowder
Assay (Titration ex Chloride)≥95.0 to ≤105.0%
Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Ludwig Bauer.; Thomas L. Welsh. The Synthesis of Derivatives of α-Mercaptoamidines1,2. J. Org. Chem. 1962, 27, (12), 4382-4385.
  2. Everett M. Stone.; Terezie H. Schaller.; Helena Bianchi.; Maria D. Person.; Walter Fast. Inactivation of Two Diverse Enzymes in the Amidinotransferase Superfamily by 2-Chloroacetamidine:  Dimethylargininase and Peptidylarginine Deiminase. Biochemistry. 1971, 44, (12), 13744-13752.