Ethyl valerate, 98%
Ethyl valerate, 98%
Ethyl valerate, 98%
Thermo Scientific Chemicals

Ethyl valerate, 98%

CAS: 539-82-2 | C7H14O2 | 130.187 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A14854.0F
also known as A14854-0F
Price (USD)
296.00
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
296.00
Each
Chemical Identifiers
CAS539-82-2
IUPAC Nameethyl pentanoate
Molecular FormulaC7H14O2
InChI KeyICMAFTSLXCXHRK-UHFFFAOYSA-N
SMILESCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.3990-1.4030 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only