2,3,5-Trimethylpyrazine, 99%
2,3,5-Trimethylpyrazine, 99%
2,3,5-Trimethylpyrazine, 99%
Thermo Scientific Chemicals

2,3,5-Trimethylpyrazine, 99%

CAS: 14667-55-1 | C7H10N2 | 122.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14870.09
also known as A14870-09
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.40
Save 5.75 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.40
Save 5.75 (15%)
Each
Chemical Identifiers
CAS14667-55-1
IUPAC Name2,3,5-trimethylpyrazine
Molecular FormulaC7H10N2
InChI KeyIAEGWXHKWJGQAZ-UHFFFAOYSA-N
SMILESCC1=CN=C(C)C(C)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5035-1.5065 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only