4-Bromo-2,6-dichloroaniline, 98%
4-Bromo-2,6-dichloroaniline, 98%
4-Bromo-2,6-dichloroaniline, 98%
4-Bromo-2,6-dichloroaniline, 98%
Thermo Scientific Chemicals

4-Bromo-2,6-dichloroaniline, 98%

CAS: 697-88-1 | C6H4BrCl2N | 240.909 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A14884.14
also known as A14884-14
Price (USD)
58.65
Online Exclusive
64.50
Save 5.85 (9%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
58.65
Online Exclusive
64.50
Save 5.85 (9%)
Each
Add to cart
Chemical Identifiers
CAS697-88-1
IUPAC Name4-bromo-2,6-dichloroaniline
Molecular FormulaC6H4BrCl2N
InChI KeyNPQBZKNXJZARBJ-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=C(Br)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)83.0-89.0?C
Appearance (Color)Cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only