(Chloromethyl)dimethylchlorosilane, 98%
(Chloromethyl)dimethylchlorosilane, 98%
(Chloromethyl)dimethylchlorosilane, 98%
Thermo Scientific Chemicals

(Chloromethyl)dimethylchlorosilane, 98%

CAS: 1719-57-9 | C3H8Cl2Si | 143.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Catalog number A14899.22
also known as A14899-22
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS1719-57-9
IUPAC Namechloro(chloromethyl)dimethylsilane
Molecular FormulaC3H8Cl2Si
InChI KeyITKVLPYNJQOCPW-UHFFFAOYSA-N
SMILESC[Si](C)(Cl)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4350-1.4390 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
CommentSpecification differs for U.S. and non-U.S. material where indicated
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only