Heptafluorobutyric acid, 99%
Heptafluorobutyric acid, 99%
Heptafluorobutyric acid, 99%
Heptafluorobutyric acid, 99%
Thermo Scientific Chemicals

Heptafluorobutyric acid, 99%

CAS: 375-22-4 | C4HF7O2 | 214.039 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14907.22
also known as A14907-22
Price (USD)
161.00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
161.00
Each
Add to cart
Chemical Identifiers
CAS375-22-4
IUPAC Nameheptafluorobutanoic acid
Molecular FormulaC4HF7O2
InChI KeyYPJUNDFVDDCYIH-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow or pale pink
FormLiquid
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Heptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.

Solubility
Miscible with water, toluene, ether, ethanol and benzene.       

Notes
Hygroscopic. Light sensitive. Incompatible withbases, oxidizing agents and reducing agents..   
RUO – Research Use Only

General References:

  1. Moreno-González, D.; Lara, F. J.; Jurgovská, N.; Gámiz-Gracia, L.; Garcia-Campana, A. M. Determination of aminoglycosides in honey by capillary electrophoresis tandem mass spectrometry and extraction with molecularly imprinted polymers. Anal. Chim. Acta 2015, 891, 321-328.
  2. Lv, J.; Chang, H.; Wang, Y.; Wang, M.; Xiao, J.; Zhang, Q.; Cheng, Y. Fluorination on polyethylenimine allows efficient 2D and 3D cell culture gene delivery. J. Mater. Chem. B 2015, 3 (4), 642-650.