Potassium trihydrogen dioxalate dihydrate, 98+%
Potassium trihydrogen dioxalate dihydrate, 98+%
Potassium trihydrogen dioxalate dihydrate, 98+%
Thermo Scientific Chemicals

Potassium trihydrogen dioxalate dihydrate, 98+%

CAS: 6100-20-5 | C4H3KO8 | 218.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A14933.0B
also known as A14933-0B
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
146.00
Save 21.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
146.00
Save 21.35
Each
Chemical Identifiers
CAS6100-20-5
IUPAC Namepotassium oxalic acid hydrogen oxalate
Molecular FormulaC4H3KO8
InChI KeyGANDVAJEIJXBQJ-UHFFFAOYSA-M
SMILES[K+].OC(=O)C(O)=O.OC(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Redox Titration)≥98.0 to ≤102.0%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Potassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. T. Honda.; K. Murase.; T. Hirato.; Y. Awakura. pH measurement in the vicinity of a cathode evolving hydrogen gas using an antimony microelectrode. Journal of Applied Electrochemistry. 1998, 28, (6), 617-622.
  2. Hong Wen.; Tonglei Li.; Kenneth R. Morris.; Kinam Park. How Solvents Affect Acetaminophen Etching Pattern Formation: Interaction between Solvent and Acetaminophen at the Solid/Liquid Interface. J. Phys. Chem. B. 2004, 108, (7), 2270-2278.