4-Chromanol, 97%
4-Chromanol, 97%
4-Chromanol, 97%
Thermo Scientific Chemicals

4-Chromanol, 97%

CAS: 1481-93-2 | C9H10O2 | 150.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A14940.03
also known as A14940-03
Price (USD)
44.20
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
44.20
Each
Chemical Identifiers
CAS1481-93-2
IUPAC Name3,4-dihydro-2H-1-benzopyran-4-ol
Molecular FormulaC9H10O2
InChI KeyMGSHXMOLUWTMGP-UHFFFAOYNA-N
SMILESOC1CCOC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream or pale yellow
Melting Point (clear melt)37.0-43.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only