Methyl propionate, 99%
Methyl propionate, 99%
Methyl propionate, 99%
Thermo Scientific Chemicals

Methyl propionate, 99%

CAS: 554-12-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number A14944.AP
also known as A14944-AP
Price (USD)
87.65
Online exclusive
96.70
Save 9.05
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
87.65
Online exclusive
96.70
Save 9.05
Each
Chemical Identifiers
CAS554-12-1
IUPAC Namemethyl propanoate
Molecular FormulaC4H8O2
InChI KeyRJUFJBKOKNCXHH-UHFFFAOYSA-N
SMILESCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Refractive Index1.3750-1.3780 @ 20?C
Assay (GC)≥98.5%
Identification (FTIR)Conforms
It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.

Solubility
Soluble in water (5g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Rose-Marie Moravie.; Jacques Corset. Conformational behaviour and vibrational spectra of methyl propionate. Chemical Physics Letters. 1974, 26, (2), 210-214.
  2. Giancarlo Albanesi.; Pietro Moggi. Methyl methacrylate by gas phase catalytic condensation of formaldehyde with methyl propionate. Applied Catalysis. 1983, 6, (3), 293-306.