Calcium acetate hydrate, 99%
Calcium acetate hydrate, 99%
Calcium acetate hydrate, 99%
Calcium acetate hydrate, 99%
Calcium acetate hydrate, 99%
Thermo Scientific Chemicals

Calcium acetate hydrate, 99%

CAS: 114460-21-8 | C4H6CaO4 | 158.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A14947.36
Price (USD)
129.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
129.00
Each
Add to cart
Chemical Identifiers
CAS114460-21-8
IUPAC Namecalcium diacetate
Molecular FormulaC4H6CaO4
InChI KeyVSGNNIFQASZAOI-UHFFFAOYSA-L
SMILES[Ca++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)<10.0%
Appearance (Color)White
FormPowder
Complexometric Titration≥98.5 to ≤101.5% (dry wt. basis)
Calcium acetate hydrate is used as a precursor to produce calcium ions in the development of calcium oxide based mesoporous carbon dioxide sorbent. It is an active component of pyroligneous acid-modified limestone. It is employed in the preparation of magnetite-calcium phosphate core-shell nanoparticles. It serves as a raw material in the preparation of hydroxyapatite-wollastonite composites. Further, it acts as a crosslinking agent in the microfluidic production of alginate hydrogel microbeads. In addition to this, it is used to induce crystallization of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium acetate hydrate is used as a precursor to produce calcium ions in the development of calcium oxide based mesoporous carbon dioxide sorbent. It is an active component of pyroligneous acid-modified limestone. It is employed in the preparation of magnetite-calcium phosphate core-shell nanoparticles. It serves as a raw material in the preparation of hydroxyapatite-wollastonite composites. Further, it acts as a crosslinking agent in the microfluidic production of alginate hydrogel microbeads. In addition to this, it is used to induce crystallization of proteins.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Peterka, T. R.; Grahek, R.; Hren, J.; Bastarda, A.; Bergles, J.; Urleb, U. Solid state compatibility study and characterization of a novel degradation product of tacrolimus in formulation. J. Pharm. Biomed. Anal. 2015, 110, 67-75.
  2. Niu, S.; Zhou, Y.; Li, H.; Lu, C.; Liu, L. An investigation on the catalytic capability of the modified white mud after activation in transesterification and kinetic calculation. Energy 2015, 89, 982-989.