4-Benzyloxybenzyl alcohol, 98+%
4-Benzyloxybenzyl alcohol, 98+%
4-Benzyloxybenzyl alcohol, 98+%
Thermo Scientific Chemicals

4-Benzyloxybenzyl alcohol, 98+%

CAS: 836-43-1 | C14H14O2 | 214.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A14949.18
also known as A14949-18
Price (USD)
129.65
Special offer
Online exclusive
Ends: 30-Jun-2026
152.00
Save 22.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
129.65
Special offer
Online exclusive
Ends: 30-Jun-2026
152.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS836-43-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Silylated GC)≥98.0%
Melting Point (clear melt)81.5-87.5?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only