2-(Trifluoromethyl)benzonitrile, 97%
2-(Trifluoromethyl)benzonitrile, 97%
2-(Trifluoromethyl)benzonitrile, 97%
Thermo Scientific Chemicals

2-(Trifluoromethyl)benzonitrile, 97%

CAS: 447-60-9 | C8H4F3N | 171.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Catalog number A14951.09
also known as A14951-09
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.70
Save 12.05 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.70
Save 12.05 (15%)
Each
Chemical Identifiers
CAS447-60-9
IUPAC Name2-(trifluoromethyl)benzonitrile
Molecular FormulaC8H4F3N
InChI KeySOZGHDCEWOLLHV-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4605-1.4655 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only