2-Chloro-4-fluorobenzoic acid, 98%
2-Chloro-4-fluorobenzoic acid, 98%
2-Chloro-4-fluorobenzoic acid, 98%
Thermo Scientific Chemicals

2-Chloro-4-fluorobenzoic acid, 98%

CAS: 2252-51-9 | C7H4ClFO2 | 174.555 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14953.06
also known as A14953-06
Price (USD)
23.65
Special offer
Online exclusive
Ends: 28-Dec-2026
27.30
Save 3.65
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
23.65
Special offer
Online exclusive
Ends: 28-Dec-2026
27.30
Save 3.65
Each
Chemical Identifiers
CAS2252-51-9
IUPAC Name2-chloro-4-fluorobenzoic acid
Molecular FormulaC7H4ClFO2
InChI KeyGRPWQLDSGNZEQE-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(F)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)181.0-188.0?C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only