Dimethyl acetone-1,3-dicarboxylate, 97%
Dimethyl acetone-1,3-dicarboxylate, 97%
Dimethyl acetone-1,3-dicarboxylate, 97%
Thermo Scientific Chemicals

Dimethyl acetone-1,3-dicarboxylate, 97%

CAS: 1830-54-2 | C7H10O5 | 174.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A14969.36
also known as A14969-36
Price (USD)
343.65
Special offer
Online exclusive
Ends: 30-Jun-2026
404.00
Save 60.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
343.65
Special offer
Online exclusive
Ends: 30-Jun-2026
404.00
Save 60.35 (15%)
Each
Chemical Identifiers
CAS1830-54-2
IUPAC Name1,5-dimethyl 3-oxopentanedioate
Molecular FormulaC7H10O5
InChI KeyRNJOKCPFLQMDEC-UHFFFAOYSA-N
SMILESCOC(=O)CC(=O)CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4415-1.4465 @ 20?C
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only