What does CorrectASE enzyme do?
The CorrectASE enzyme has the ability to remove mismatches caused by oligonucleotide synthesis errors and is typically used in a do-it-yourself gene synthesis workflow to aid in decreasing mutations in your synthetic genes/fragements.