2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.
Solubility
Soluble in water (0.018g/L).
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
Geoffrey G. Briggs, J. E. Dawson. Hydrolysis of 2,6-dichlorobenzonitrile in soils. J. Agric. Food Chem. 1970, 18, (1), 97-99.