1-Dodecene, 96%
1-Dodecene, 96%
1-Dodecene, 96%
Thermo Scientific Chemicals

1-Dodecene, 96%

CAS: 112-41-4 | C12H24 | 168.324 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A14992.22
also known as A14992-22
Price (USD)
77.20
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
77.20
Each
Chemical Identifiers
CAS112-41-4
IUPAC Namedodec-1-ene
Molecular FormulaC12H24
InChI KeyCRSBERNSMYQZNG-UHFFFAOYSA-N
SMILESCCCCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥95.0%
Refractive Index1.4270-1.4330 @ 20?C
FormLiquid
1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.

Solubility
It is soluble in ethanol, ethyl ether, and acetone

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
RUO – Research Use Only

General References:

  1. Kazuhiko Sato.; Masao Aoki.; Masami Ogawa.; Tadashi Hashimoto.; Ryoji Noyori. A Practical Method for Epoxidation of Terminal Olefins with 30% Hydrogen Peroxide under Halide-Free Conditions. J. Org. Chem. 1996, 61, (23), 8310-8311.
  2. Christine E. Garrett.; Gregory C. Fu. Hydroboration of Olefins with Catecholborane at Room Temperature in the Presence of N,N-Dimethylacetamide. J. Org. Chem. 1996, 61, (10), 3224-3225.