Ethyl levulinate, 98%
Ethyl levulinate, 98%
Ethyl levulinate, 98%
Thermo Scientific Chemicals

Ethyl levulinate, 98%

CAS: 539-88-8 | C7H12O3 | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Catalog number A15001.36
also known as A15001-36
Price (USD)
138.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
138.00
Each
Chemical Identifiers
CAS539-88-8
IUPAC Nameethyl 4-oxopentanoate
Molecular FormulaC7H12O3
InChI KeyGMEONFUTDYJSNV-UHFFFAOYSA-N
SMILESCCOC(=O)CCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.4200-1.4240 @ 20?C
Assay (GC)≥97.5%
Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Shunmugavel Saravanamurugan.; Anders Riisager. Solid acid catalysed formation of ethyl levulinate and ethyl glucopyranoside from mono- and disaccharides. Catalysis Communications. 2012, 7, 71-75.
  2. Chun Chang.; Guizhuan Xu.; Xiaoxian Jiang. Production of ethyl levulinate by direct conversion of wheat straw in ethanol media. Bioresource Technology. 2012, 121, 93-99.