Methyl phenyl sulfoxide, 98+%
Methyl phenyl sulfoxide, 98+%
Methyl phenyl sulfoxide, 98+%
Thermo Scientific Chemicals

Methyl phenyl sulfoxide, 98+%

CAS: 1193-82-4 | C7H8OS | 140.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15009.06
also known as A15009-06
Price (USD)
44.65
Online Exclusive
49.20
Save 4.55 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
44.65
Online Exclusive
49.20
Save 4.55 (9%)
Each
Chemical Identifiers
CAS1193-82-4
IUPAC Namemethanesulfinylbenzene
Molecular FormulaC7H8OS
InChI KeyJXTGICXCHWMCPM-UHFFFAOYNA-N
SMILESCS(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Water Content (Karl Fischer Titration)≤0.5%
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥98.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only