2,6-Dimethylbenzoic acid, 98+%
2,6-Dimethylbenzoic acid, 98+%
2,6-Dimethylbenzoic acid, 98+%
2,6-Dimethylbenzoic acid, 98+%
Thermo Scientific Chemicals

2,6-Dimethylbenzoic acid, 98+%

CAS: 632-46-2 | C9H10O2 | 150.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A15014.03
also known as A15014-03
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.00
Save 5.35 (14%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.00
Save 5.35 (14%)
Each
Add to cart
Chemical Identifiers
CAS632-46-2
IUPAC Name2,6-dimethylbenzoic acid
Molecular FormulaC9H10O2
InChI KeyHCBHQDKBSKYGCK-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Melting Point (clear melt)113-118?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only