7-Amino-4-methylcoumarin, 98%
7-Amino-4-methylcoumarin, 98%
7-Amino-4-methylcoumarin, 98%
Thermo Scientific Chemicals

7-Amino-4-methylcoumarin, 98%

CAS: 26093-31-2 | C10H9NO2 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number A15017.06
also known as A15017-06
Price (USD)
441.65
Online Exclusive
491.00
Save 49.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
441.65
Online Exclusive
491.00
Save 49.35 (10%)
Each
Chemical Identifiers
CAS26093-31-2
IUPAC Name7-amino-4-methyl-2H-chromen-2-one
Molecular FormulaC10H9NO2
InChI KeyGLNDAGDHSLMOKX-UHFFFAOYSA-N
SMILESCC1=CC(=O)OC2=CC(N)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
FormPowder or crystalline powder
Assay (GC)≥97.5%
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
View more
Used as matrix for analysis of monosulfated disaccharides and as a co-matrix with 6-aza-2-thiothymidine for sulfated neutral and salivated tri-and tetrasaccharides. Reagent for preparing new fluorogenic AMC-based substrates for cystine amino peptidase (and other hydrolases). Also used as reference compound in the enzyme assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as matrix for analysis of monosulfated disaccharides and as a co-matrix with 6-aza-2-thiothymidine for sulfated neutral and salivated tri-and tetrasaccharides. Reagent for preparing new fluorogenic AMC-based substrates for cystine amino peptidase (and other hydrolases). Also used as reference compound in the enzyme assay.

Solubility
Soluble in acetone (10 mg/ml) dimethyl sulfoxide (10 mg/ml) and dimethylformamide.

Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. M Zimmerman; B Ashe; E C Yurewicz; G Patel; M. Zimmerman; B. Ashe; E.C. Yurewicz; G. Patel. Sensitive assays for trypsin, elastase, and chymotrypsin using new fluorogenic substrates. Analytical Biochemistry. 1977, 78 (1), 47-51.
  2. Yuqin Dai et. al. Matrix-assisted laser desorption ionization mass spectrometry for the analysis of monosulfated oligosaccharides. Carbohydrate Research. 1997, 304 (1), 1-9.