Ethylamine, 70% aq. soln.
Ethylamine, 70% aq. soln.
Thermo Scientific Chemicals

Ethylamine, 70% aq. soln.

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A15022.AP
also known as A15022-AP
Price (USD)
29.20
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
29.20
Each
Add to cart
Chemical Identifiers
CAS75-04-7
IUPAC Nameethanamine
Molecular FormulaC2H7N
InChI KeyQUSNBJAOOMFDIB-UHFFFAOYSA-N
SMILESCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Assay (unspecified)68-72% w/w Ethylamine content
Refractive Index1.3815-1.3865 @ 20?C

Ethylamine 70% aq. solution is widely used in chemical industry for the synthesis of diethyldiazene, dimethylolethyltriazone, ethylcyanopyrolidone disperse and 1,3-diethylthiourea. It is also used in the production of mining chemicals and flotation agents such as isopropylethyl thiocarbamates. In polymer industry, it is used as a chain stopper in polyurethane production to control degree of polymerization. It is a precursor used for the preparation of benzilnitrate, detergents, rayon, rocket propellant, and in preparation of alkyl isocyanates, which find application in the manufacture of pharmaceuticals. In rubber industry, it is utilized as a vulcanization accelerator for sulfur cured rubbers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.