1-Chloronaphthalene, tech. 85%, remainder 2-chloronaphthalene
1-Chloronaphthalene, tech. 85%, remainder 2-chloronaphthalene
1-Chloronaphthalene, tech. 85%, remainder 2-chloronaphthalene
Thermo Scientific Chemicals

1-Chloronaphthalene, tech. 85%, remainder 2-chloronaphthalene

CAS: 90-13-1 | C10H7Cl | 162.616 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A15029.30
also known as A15029-30
Price (USD)
81.65
Online Exclusive
90.50
Save 8.85 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
81.65
Online Exclusive
90.50
Save 8.85 (10%)
Each
Chemical Identifiers
CAS90-13-1
IUPAC Name1-chloronaphthalene
Molecular FormulaC10H7Cl
InChI KeyJTPNRXUCIXHOKM-UHFFFAOYSA-N
SMILESClC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.6290-1.6370 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥83.0% (major isomer)
1-Chloronaphthalene was used as solvent additive in the fabrication of fullerene based photovoltaic devices. It was used in preparation of polyvinyl chloride based membranes of 5,10,15,20-tetrakis(4-methoxyphenyl)porphyrinatocobalt(II) for arsenite determination. Also used in the synthesis of pharmaceutical compounds such as aryl chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloronaphthalene was used as solvent additive in the fabrication of fullerene based photovoltaic devices. It was used in preparation of polyvinyl chloride based membranes of 5,10,15,20-tetrakis(4-methoxyphenyl)porphyrinatocobalt(II) for arsenite determination. Also used in the synthesis of pharmaceutical compounds such as aryl chlorides.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Gisela L Schulz et. al. Optimization of solution-processed oligothiophene:fullerene based organic solar cells by using solvent additives. Beilstein Journal of Nanotechnology,. 2013, 4 680-689.
  2. V.K. Gupta; and S. Agarwal. PVC based 5,10,15,20-tetrakis (4-methoxyphenyl) porphyrinatocobalt(II) membrane potentiometric sensor for arsenite. Talanta. 2005, 65 (3), 730-734 .