L-Thiazolidine-4-carboxylic acid, 98%
L-Thiazolidine-4-carboxylic acid, 98%
L-Thiazolidine-4-carboxylic acid, 98%
Thermo Scientific Chemicals

L-Thiazolidine-4-carboxylic acid, 98%

CAS: 34592-47-7 | C4H7NO2S | 133.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A15033.09
also known as A15033-09
Price (USD)
30.60
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
30.60
Each
Chemical Identifiers
CAS34592-47-7
IUPAC Name(4R)-1,3-thiazolidin-3-ium-4-carboxylate
Molecular FormulaC4H7NO2S
InChI KeyDZLNHFMRPBPULJ-VKHMYHEASA-N
SMILES[O-]C(=O)[C@@H]1CSC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Optical Rotation-138° to -144° (c=1 in water)
FormCrystals or powder or crystalline powder or needles
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only