2-Iodopropane, 98+%, stab. with copper
2-Iodopropane, 98+%, stab. with copper
2-Iodopropane, 98+%, stab. with copper
Thermo Scientific Chemicals

2-Iodopropane, 98+%, stab. with copper

CAS: 75-30-9 | C3H7I | 169.993 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A15066.14
also known as A15066-14
Price (USD)
35.30
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
35.30
Each
Chemical Identifiers
CAS75-30-9
IUPAC Name2-iodopropane
Molecular FormulaC3H7I
InChI KeyFMKOJHQHASLBPH-UHFFFAOYSA-N
SMILESCC(C)I
View more
SpecificationsSpecification SheetSpecification Sheet
StabilizerCopper powder
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Refractive Index1.4960-1.5000 @ 20?C
Assay (GC)≥98.0%
View more
2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

Solubility
Soluble in water(1.4g/L).

Notes
Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. The ring-opening of cyclic thionocarbonates followed by reductive elimination with zinc to give alkenes provides an alternative to the Corey-Winter desulfurization method (see Triethyl phosphite, L00339): Tetrahedron Lett., 3793 (1973).