2-Bromobutane, 98%
2-Bromobutane, 98%
2-Bromobutane, 98%
Thermo Scientific Chemicals

2-Bromobutane, 98%

CAS: 78-76-2 | C4H9Br | 137.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A15069.30
also known as A15069-30
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
55.90
Save 8.25 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
55.90
Save 8.25 (15%)
Each
Chemical Identifiers
CAS78-76-2
IUPAC Name2-bromobutane
Molecular FormulaC4H9Br
InChI KeyUPSXAPQYNGXVBF-UHFFFAOYNA-N
SMILESCCC(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4350-1.4390 @ 20°C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
2-Bromobutane is utilized in the synthesis of non-nucleosides. It is used as a solvent as well as an intermediate in organic synthesis. It is an important raw material used to prepare Grignard reagent by reacting with magnesium. Further, it reacts with hydrocyanic acid to prepare sodium salt to get (+-)-sec-butyl cyanide. It is an important raw material used to prepare Grignard reagent by reacting with magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobutane is utilized in the synthesis of non-nucleosides. It is used as a solvent as well as an intermediate in organic synthesis. It is an important raw material used to prepare Grignard reagent by reacting with magnesium. Further, it reacts with hydrocyanic acid to prepare sodium salt to get (+-)-sec-butyl cyanide. It is an important raw material used to prepare Grignard reagent by reacting with magnesium.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, magnesium, sodium, sodium oxides and potassium.
RUO – Research Use Only

General References:

  1. Vatsal, A.; Zinjarde, S. S.; Kumar, A. R. Yarrowia lipolytica NCIM 3589, a tropical marine yeast, degrades bromoalkanes by an initial hydrolytic dehalogenation step. Biodegradation 2015, 26 (2), 127-138.
  2. Sannasi, V.; Jose, T. P.; Jeyakumar, D. Synthesis and optical properties of poly(2,7-(9,9-dihexylfluorene)-3,3'(4, 4'-dialkoxybiphenyl)). Des. Monomers Polym. 2015, 18 (1), 51-63.