3-(4-Carboxyphenyl)propionic acid, 98%
3-(4-Carboxyphenyl)propionic acid, 98%
3-(4-Carboxyphenyl)propionic acid, 98%
Thermo Scientific Chemicals

3-(4-Carboxyphenyl)propionic acid, 98%

CAS: 38628-51-2 | C10H10O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
100 g
Catalog number A15081.09
also known as A15081-09
Price (USD)
158.65
Online Exclusive
176.00
Save 17.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
158.65
Online Exclusive
176.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS38628-51-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Melting Point285-292°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only