2-Chloro-6-fluorobenzylideneacetone, 98%
2-Chloro-6-fluorobenzylideneacetone, 98%
2-Chloro-6-fluorobenzylideneacetone, 98%
Thermo Scientific Chemicals

2-Chloro-6-fluorobenzylideneacetone, 98%

CAS: 175136-82-0 | C10H8ClFO | 198.621 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A15083.03
also known as A15083-03
Price (USD)
27.80
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
27.80
Each
Chemical Identifiers
CAS175136-82-0
IUPAC Name(3E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one
Molecular FormulaC10H8ClFO
InChI KeyVZRBDBXFAKDNDJ-AATRIKPKSA-N
SMILESCC(=O)C=CC1=C(F)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Assay (GC)≥97.5%
Refractive Index1.5770-1.5810 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only