4-Bromoaniline, 98+%
4-Bromoaniline, 98+%
4-Bromoaniline, 98+%
Thermo Scientific Chemicals

4-Bromoaniline, 98+%

CAS: 106-40-1 | C6H6BrN | 172.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A15126.0B
also known as A15126-0B
Price (USD)
357.65
Special offer
Online exclusive
Ends: 30-Jun-2026
420.00
Save 62.35 (15%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
357.65
Special offer
Online exclusive
Ends: 30-Jun-2026
420.00
Save 62.35 (15%)
Each
Chemical Identifiers
CAS106-40-1
IUPAC Name4-bromoaniline
Molecular FormulaC6H6BrN
InChI KeyWDFQBORIUYODSI-UHFFFAOYSA-N
SMILESNC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)Pale cream to cream to yellow to brown to gray to black
FormCrystals or powder or crystalline powder or lumps or fused solid
Identification (FTIR)Conforms
Melting Point (clear melt)59.0-65.0?C
4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.

Solubility
Soluble in alcohol and ether. Insoluble in cold water.

Notes
Store in a cool place. Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Gao, W. Synthesis and Structure Analysis of N-diphenylphosphoryl-4-bromoaniline. Mol. Cryst. Liq. Cryst. 2015, 609 (1), 155-160.
  2. Štrukil, V.; Gracin, D.; Magdysyuk, O. V.; Dinnebier, R. E.; Friščić, T. Trapping Reactive Intermediates by Mechanochemistry: Elusive Aryl N-Thiocarbamoylbenzotriazoles as Bench-Stable Reagents. Angew. Chem. Int. Ed. 2015, 54 (29), 8440-8443.