1-Bromohexadecane, 98%
1-Bromohexadecane, 98%
1-Bromohexadecane, 98%
Thermo Scientific Chemicals

1-Bromohexadecane, 98%

CAS: 112-82-3 | C16H33Br | 305.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1000 g
Catalog number A15147.14
also known as A15147-14
Price (USD)
25.70
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
25.70
Each
Chemical Identifiers
CAS112-82-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to brown
Assay (GC)≥97.5%
Refractive Index1.4605-1.4645 @ 20?C
FormFused solid or clear liquid as melt
Identification (FTIR)Conforms
1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Pretima Manoj.; Andrew D Watson.; David J Hibberd.; Annette J Fillery-Travis.; Margaret M Robins. II. Steady-State Rheological Investigations ☆. Journal of Colloid and Interface Science. 1998, 207, (2), 294-302.
  2. Domenico Albanese.; Dario Landini.; Michele Penso. Hydrated Tetrabutylammonium Fluoride as a Powerful Nucleophilic Fluorinating Agent. J. Org. Chem. 1998, 63, (25), 9587-9589.