2-Isopropylnaphthalene, 96%
2-Isopropylnaphthalene, 96%
2-Isopropylnaphthalene, 96%
Thermo Scientific Chemicals

2-Isopropylnaphthalene, 96%

CAS: 2027-17-0 | C13H14 | 170.255 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15153.06
also known as A15153-06
Price (USD)
74.65
Special offer
Online exclusive
Ends: 28-Dec-2026
87.70
Save 13.05
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 28-Dec-2026
87.70
Save 13.05
Each
Chemical Identifiers
CAS2027-17-0
IUPAC Name2-(propan-2-yl)naphthalene
Molecular FormulaC13H14
InChI KeyTVYVQNHYIHAJTD-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
FormLiquid
Refractive Index1.5840-1.5900 @ 20?C
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only