SensiCell™ DMEM/F12 - FAQs
SensiCell™ DMEM/F12 - FAQs
View additional product information for SensiCell™ DMEM/F12 - FAQs (A1515501)
1 product FAQs found
What alternative do you offer for the discontinued SensiCell DMEM/F-12?
The alternative product is Advanced DMEM/F-12 (Cat. No. 12634-010). Please add 2.5 mM GlutaMAX Supplement to the medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.