SensiCell™ MEM - FAQs
SensiCell™ MEM - FAQs
View additional product information for SensiCell™ MEM - FAQs (A1515601)
1 product FAQs found
What alternative do you offer for the discontinued SensiCell MEM?
The alternative product is Advanced MEM (Cat. No. 12492-013). Please add 2 mM GlutaMAX Supplement to the medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.