SensiCell™ RPMI 1640 Medium - FAQs
SensiCell™ RPMI 1640 Medium - FAQs
View additional product information for SensiCell™ RPMI 1640 Medium - FAQs (A1515701)
1 product FAQs found
What alternative do you offer for the discontinued SensiCell RPMI 1640?
The alternative product is Advanced RPMI 1640 (Cat. No. 12633-012). Please add 2 mM GlutaMAX Supplement to the medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.