Phenylpropiolic acid, 98+%
Phenylpropiolic acid, 98+%
Phenylpropiolic acid, 98+%
Thermo Scientific Chemicals

Phenylpropiolic acid, 98+%

CAS: 637-44-5 | C9H6O2 | 146.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
Catalog number A15163.09
also known as A15163-09
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS637-44-5
IUPAC Name3-phenylprop-2-ynoic acid
Molecular FormulaC9H6O2
InChI KeyXNERWVPQCYSMLC-UHFFFAOYSA-N
SMILESOC(=O)C#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.0%
View more
Phenylpropiolic acid is used as a Pharmaceutical Intermediates, laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylpropiolic acid is used as a Pharmaceutical Intermediates, laboratory chemicals.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Leif Schweitz.; Lars I. Andersson.; Staffan Nilsson. Molecular imprint-based stationary phases for capillary electrochromatography. Journal of Chromatography A. 1998, 817, (1-2), 5-13.
  2. Arti Arora.; Muraleedharan G. Nair.; Gale M. Strasburg. Structure-Activity Relationships for Antioxidant Activities of a Series of Flavonoids in a Liposomal System. Free Radical Biology and Medicine. 1998, 24, (9), 1355-1363.