3,4-Dichloroaniline, 98%
3,4-Dichloroaniline, 98%
3,4-Dichloroaniline, 98%
3,4-Dichloroaniline, 98%
Thermo Scientific Chemicals

3,4-Dichloroaniline, 98%

CAS: 95-76-1 | C6H5Cl2N | 162.013 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
500 g
2500 g
Catalog number A15199.18
also known as A15199-18
Price (USD)
27.90
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
27.90
Each
Add to cart
Chemical Identifiers
CAS95-76-1
IUPAC Name3,4-dichloroaniline
Molecular FormulaC6H5Cl2N
InChI KeySDYWXFYBZPNOFX-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or brown or grey containing some darker particles
FormCrystals or crystalline lumps or powder or crystalline powder of flakes
Assay (GC)≥97.5%
CommentDarker particles are coarser crystalline lumps
CommentMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only