Pamoic acid, 99%
Pamoic acid, 99%
Pamoic acid, 99%
Thermo Scientific Chemicals

Pamoic acid, 99%

CAS: 130-85-8 | C23H16O6 | 388.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A15207.18
also known as A15207-18
Price (USD)
57.65
Online Exclusive
63.80
Save 6.15 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
63.80
Save 6.15 (10%)
Each
Chemical Identifiers
CAS130-85-8
IUPAC Name4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
Molecular FormulaC23H16O6
InChI KeyWLJNZVDCPSBLRP-UHFFFAOYSA-N
SMILESOC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)Yellow to yellow-green
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.

Solubility
Soluble in nitrobenzene, pyridine

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Kristof Prinsen.; Junjie Li.; Hubert Vanbilloen.; Peter Vermaelen.; Ellen Devos.; Luc Mortelmans.; Guy Bormans.; Yicheng Ni.; Alfons Verbruggen. Development and evaluation of a 68Ga labeled pamoic acid derivative for in vivo visualization of necrosis using positron emission tomography. Bioorganic & Medicinal Chemistry. 2010, 18, (14), 5274-5281.
  2. Miao Du.; Zhi-Hui Zhang.; Wei Guo.; Xiao-Juan Fu. Multi-Component Hydrogen-Bonding Assembly of a Pharmaceutical Agent Pamoic Acid with Piperazine or 4,4'-Bipyridyl: A Channel Hydrated Salt with Multiple-Helical Motifs vs a Bimolecular Cocrystal. Cryst. Growth Des. 2009, 9, (4), 1655-1657.