5-Chlorothiophene-2-carboxylic acid, 98%
5-Chlorothiophene-2-carboxylic acid, 98%
5-Chlorothiophene-2-carboxylic acid, 98%
5-Chlorothiophene-2-carboxylic acid, 98%
Thermo Scientific Chemicals

5-Chlorothiophene-2-carboxylic acid, 98%

CAS: 24065-33-6 | C5H3ClO2S | 162.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15209.14
also known as A15209-14
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS24065-33-6
IUPAC Name5-chlorothiophene-2-carboxylic acid
Molecular FormulaC5H3ClO2S
InChI KeyQZLSBOVWPHXCLT-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(Cl)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)146.0-153.0?C
Appearance (Color)White to cream to yellow
FormPowder
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only