2,3'-Dibromoacetophenone, 97%
2,3'-Dibromoacetophenone, 97%
2,3'-Dibromoacetophenone, 97%
Thermo Scientific Chemicals

2,3'-Dibromoacetophenone, 97%

CAS: 18523-22-3 | C8H6Br2O | 277.943 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A15210.14
also known as A15210-14
Price (USD)
246.65
Online Exclusive
274.00
Save 27.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
246.65
Online Exclusive
274.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS18523-22-3
IUPAC Name2-bromo-1-(3-bromophenyl)ethan-1-one
Molecular FormulaC8H6Br2O
InChI KeyMZBXSQBQPJWECM-UHFFFAOYSA-N
SMILESBrCC(=O)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)45.0-55.0?C
It is employed as an intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate for pharmaceutical and organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Utpalendu S. Maitra; and Helmut Ankel. Uridine Diphosphate-4-Keto-Glucose, an Intermediate in the Uridine Diphosphate-Galactose-4-Epimerase Reaction. Current Issue, 1971, 68(11), 2660-2663.
  2. Eduardo L. Setti, et al. Design and synthesis of tetracyclic nonpeptidic biaryl nitrile inhibitors of cathepsin K. Bioorganic & Medicinal Chemistry Letters, 2006, 16(16), 4296-4299.