2-Methoxyphenylacetic acid, 99%
2-Methoxyphenylacetic acid, 99%
2-Methoxyphenylacetic acid, 99%
Thermo Scientific Chemicals

2-Methoxyphenylacetic acid, 99%

CAS: 93-25-4 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
Catalog number A15211.09
also known as A15211-09
Price (USD)
29.65
Online exclusive
33.30
Save 3.65
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
29.65
Online exclusive
33.30
Save 3.65
Each
Chemical Identifiers
CAS93-25-4
IUPAC Name2-(2-methoxyphenyl)acetic acid
Molecular FormulaC9H10O3
InChI KeyIVEWTCACRDEAOB-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.5%
Melting Point (clear melt)121.5-126.5?C
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5% to ≤101.5%
View more
2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water. Also soluble in methanol, ether, ethanol and acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Hans-Adolf Arfmann; Wolf-Rainer Abraham. Microbial reduction of aromatic carboxylic acids. Zeitschrift fur Naturforschung C. 1993, 48,(1-2), 52-57.
  2. H.Pietilainena; T.Saesmaaab. HPLC determination of pindolol benzoate and pindolol 2-methoxyphenylacetate. Journal of Liquid Chromatography & Related Technologies. 1996, 19,(4), 583-591.