9-Fluorenylmethanol, 99%
9-Fluorenylmethanol, 99%
9-Fluorenylmethanol, 99%
Thermo Scientific Chemicals

9-Fluorenylmethanol, 99%

CAS: 24324-17-2 | C14H12O | 196.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15212.14
also known as A15212-14
Price (USD)
178.65
Special offer
Online exclusive
Ends: 30-Jun-2026
210.00
Save 31.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
178.65
Special offer
Online exclusive
Ends: 30-Jun-2026
210.00
Save 31.35 (15%)
Each
Chemical Identifiers
CAS24324-17-2
IUPAC Name(9H-fluoren-9-yl)methanol
Molecular FormulaC14H12O
InChI KeyXXSCONYSQQLHTH-UHFFFAOYSA-N
SMILESOCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)102.0-108.0?C
Assay (GC)≥98.5%
9-Fluorenylmethanol acts as a N-protecting reagent, which is used in the synthesis of peptide. It is also used in the preparation of deoxynucleoside 9-fluorenemethyl phosphorodithioates. Further, it is used to prepare 9-(fluoromethyl)fluorene. In addition to this, it is involved in the electropolymerization with boron trifluoride diethyl etherate to yield low-potential electrodeposition of semiconducting poly(9-fluorenemethanol) film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Fluorenylmethanol acts as a N-protecting reagent, which is used in the synthesis of peptide. It is also used in the preparation of deoxynucleoside 9-fluorenemethyl phosphorodithioates. Further, it is used to prepare 9-(fluoromethyl)fluorene. In addition to this, it is involved in the electropolymerization with boron trifluoride diethyl etherate to yield low-potential electrodeposition of semiconducting poly(9-fluorenemethanol) film.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Reagent for protection of carboxyl groups as their 9-fluorenylmethyl (Fm) esters, which improve solubility of the molecule in organic solvents. Fm esters can be formed, e.g. by the DCC-DMAP method: Tetrahedron Lett., 24, 281 (1983), or by imidazole-catalyzed transesterification of active esters of N-protected amino acids: Int. J. Pept. Prot. Res., 21, 196 (1983). Cleavage is by hydrogenolysis. See Appendix 6.
  2. Can also be used to protect phosphate groups in oligonucleotide synthesis: J. Chem. Soc., Chem. Commun., 803 (1973).
  3. Arimitsu, K.; Kumazawa, Y.; Furutani, M. Preparation of base-amplifying microcapsules and their application to photoreactive materials. J. Polym. Sci., Part A: Polym. Chem. 2015, 53 (21), 2440-2443.
  4. Furutani, M.; Kobayashi, H.; Gunji, T.; Abe, Y.; Arimitsu, K. Base-amplifying silicone resins with photobase-generating side chains and their application to negative-working photoresists. J. Polym. Sci., Part A: Polym. Chem. 2015, 53 (10), 1205-1212.