Tetra-n-heptylammonium bromide, 99%
Tetra-n-heptylammonium bromide, 99%
Tetra-n-heptylammonium bromide, 99%
Thermo Scientific Chemicals

Tetra-n-heptylammonium bromide, 99%

CAS: 4368-51-8 | C28H60BrN | 490.699 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15218.06
also known as A15218-06
Price (USD)
33.65
Online Exclusive
37.30
Save 3.65 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
33.65
Online Exclusive
37.30
Save 3.65 (10%)
Each
Chemical Identifiers
CAS4368-51-8
IUPAC Nametetraheptylazanium bromide
Molecular FormulaC28H60BrN
InChI KeyYQIVQBMEBZGFBY-UHFFFAOYSA-M
SMILES[Br-].CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or flakes
Assay (Titration ex Bromide)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤0.5%
Melting Point85.0-92.0?C
View more
Tetra-n-heptylammonium bromide is used as a mobile phase for the separation of a series of glucosinolates by reversed-phase ion-pair liquid chromatography. It acts as an ion pair reagent, which is used in the extraction of molybdenum complexes into toluene film. Further, it is used in organic synthesis. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-heptylammonium bromide is used as a mobile phase for the separation of a series of glucosinolates by reversed-phase ion-pair liquid chromatography. It acts as an ion pair reagent, which is used in the extraction of molybdenum complexes into toluene film. Further, it is used in organic synthesis. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kumar, A. S. K.; Jiang, S. J.; Tseng, W. L. Effective adsorption of chromium(vi)/Cr(iii) from aqueous solution using ionic liquid functionalized multiwalled carbon nanotubes as a super sorbent. J. Mater. Chem. A 2015, 3 (13), 7044-7057.
  2. Takahashi, M.; Sakurai, K.; Fujii, H.; Saito, K. Identification of Indicator Components for the Discrimination of Cassia Plants in Health Teas and Development of Analytical Method for the Components. J. AOAC Int. 2014, 97 (4), 1195-1201.